Anti-HSPB1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24533
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 27kDa protein 1
- Antigen symbol:HSPB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:3315
- Antigen synonyms:SRP27|HSP27|HSP28|Hsp25|HEL-S-102|HS.76067|HMN2B|CMT2F
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: