Anti-PIF1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The PIF1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIF1. This antibody reacts with mouse. The PIF1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC / IF (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:300
Type: Primary
Antigen: PIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- NOVUNBP1-97931
- NOVUNBP1-97931SS
- Antigen symbol:
- PIF1
- PIF1
- Antigen synonyms:
- PIF|C15orf20
- PIF|C15orf20
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 80119
- 80119
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together