Anti-LC3I Rabbit Polyclonal Antibody
Rabbit Polyclonal LC3I Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Primate, Porcine.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: LC3I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
- Catalog No:
- NOVUNBP1-78964SS
- NOVUNBP1-78964
- Antigen symbol:
- LC3I
- LC3I
- Antigen name:
- microtubule-associated protein 1 light chain 3 alpha
- microtubule-associated protein 1 light chain 3 alpha
- Antigen synonyms:
- MAP1BLC3|MAP1ALC3|LC3A|ATG8E|LC3
- MAP1BLC3|MAP1ALC3|LC3A|ATG8E|LC3
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 84557
- 84557
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together