Anti-RUNX2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The RUNX2 / CBFA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RUNX2 / CBFA1. This antibody reacts with human, mouse. The RUNX2 / CBFA1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: RUNX2/CBFA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-77461SS
- NOVUNBP1-77461
- Antigen symbol:
- RUNX2
- RUNX2
- Antigen name:
- Runt-related Transcription Factor 2
- Runt-related Transcription Factor 2
- Antigen synonyms:
- Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
- Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 860
- 860
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together