Anti-TGOLN2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The TGN46 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TGN46. This antibody reacts with human. The TGN46 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: TGN46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP1-49643
- NOVUNBP1-49643SS
- Antigen symbol:
- TGOLN2
- TGOLN2
- Antigen name:
- Trans-Golgi Network Integral Membrane Protein 2
- Trans-Golgi Network Integral Membrane Protein 2
- Antigen synonyms:
- TGN38|trans-golgi network protein 2|TGN51|TGN48|TTGN2|TGN46
- TGN38|trans-golgi network protein 2|TGN51|TGN48|TTGN2|TGN46
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 10618
- 10618
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together