Anti-N Cadherin Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 13A9]
Catalog # NOVUNBP1-48309F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:cadherin 2, type 1, N-cadherin (neuronal)
- Antigen symbol:CDH2
- Clonality:Monoclonal
- Clone:13A9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:1000
- Antigen synonyms:CDHN|CD325|CDw325|NCAD
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: N Cadherin
Clonality: monoclonal
Clone: 13A9
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat