Anti-HPd1 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: DHIC2-4A10]
Catalog # NOVUNBP1-18953PCP
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:HPD1
- Clonality:Monoclonal
- Clone:DHIC2-4A10
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: HPd1
Clonality: monoclonal
Clone: DHIC2-4A10
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human