Anti-AGR2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-05936F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:anterior gradient 2, protein disulphide isomerase family member
- Antigen symbol:AGR2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:10551
- Antigen synonyms:AG2|HAG-2|PDIA17|XAG-2|GOB-4|HEL-S-116
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: AGR2 (anterior gradient 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: