Anti-PDE9A Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-00641C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Phosphodiesterase 9A
- Antigen symbol:PDE9A
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:5152
- Antigen synonyms:HSPDE9A2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PDE9A (phosphodiesterase 9A)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat