Anti-MLXIPL Rabbit Polyclonal Antibody (DyLight® 488)
NOVUNB400-136G
- Antibody type:Primary
- Antigen name:Carbohydrate-responsive element-binding protein
- Antigen symbol:ChREBP
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:51085
- Antigen synonyms:WBSCR14|WS-bHLH|MIO|MONDOB|Williams Beuren Syndrome Chromosome Region 14
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: MLXIPL (MLX interacting protein-like)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse