Order Entry
France
ContactUsLinkComponent
 
Anti-MLXIPL Rabbit Polyclonal Antibody (Biotin)
  NOVUNB400-135B
Anti-MLXIPL Rabbit Polyclonal Antibody (Biotin)
  NOVUNB400-135B

 

  • Antibody type:
    Primary
  • Antigen name:
    Carbohydrate-responsive element-binding protein
  • Antigen symbol:
    ChREBP
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    51085
  • Antigen synonyms:
    WBSCR14|WS-bHLH|MIO|MONDOB|Williams Beuren Syndrome Chromosome Region 14
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: MLXIPL (MLX interacting protein-like)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat