Order Entry
France
ContactUsLinkComponent
Anti-SCARB2 Rabbit Polyclonal Antibody
Anti-SCARB2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SCARB2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB400-102
  • NOVUNB400-102SS
  • Antigen symbol:
  • SCARB2
  • SCARB2
  • Antigen name:
  • Scavenger receptor class B, member 2
  • Scavenger receptor class B, member 2
  • Antigen synonyms:
  • LIMP-2|EPM4|CD36L2|AMRF|LIMPII|SR-BII|LGP85|HLGP85
  • LIMP-2|EPM4|CD36L2|AMRF|LIMPII|SR-BII|LGP85|HLGP85
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 950
  • 950
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together