Anti-SLC7A11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB300-318H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 7 Member 11
- Antigen symbol:Solute carrier family 7, member 11
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:23657
- Antigen synonyms:Xc- System)|Amino Acid Transport System Xc-|Member 11|CCBR1,Calcium Channel Blocker Resistance Protein|Solute Carrier Family 7 (Anionic Amino Acid Transporter Light Chain
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat