Order Entry
France
ContactUsLinkComponent
Anti-SLC7A11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-SLC7A11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB300-318F
Supplier:  Novus Biologicals
Anti-SLC7A11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB300-318F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11
  • Antigen symbol:
    Solute carrier family 7, member 11
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    23657
  • Antigen synonyms:
    xCT|CCBR1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat