Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27a]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: NF1 (neurofibromin 1)
Clonality: monoclonal
Clone: McNFn27a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Guinea pig, Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB300-153SS
- NOVUNB300-153
- Antigen symbol:
- NF1
- NF1
- Antigen name:
- Neurofibromin 1
- Neurofibromin 1
- Antigen synonyms:
- Neurofibromin|NF1|VRNF|NFNS|WSS|neurofibromin|DKFZp686J1293
- Neurofibromin|NF1|VRNF|NFNS|WSS|neurofibromin|DKFZp686J1293
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- McNFn27a
- McNFn27a
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4763
- 4763
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together