Anti-DRD1 Mouse Monoclonal Antibody (DyLight® 650) [clone: SG2-D1a]
Catalog # NOVUNB110-60017C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Dopamine receptor D1
- Antigen symbol:DRD1
- Clonality:Monoclonal
- Clone:SG2-D1a
- Conjugation:DyLight® 650
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:1812
- Antigen synonyms:DRD1A|DADR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: DRD1 (dopamine receptor D1)
Clonality: monoclonal
Clone: SG2-D1a
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Mouse, Rat