Anti-CD36 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: D-2712]
Catalog # NOVUNB110-59724F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:CD36 molecule (thrombospondin receptor)
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:D-2712
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgA kappa
- Reactivity:Rat,Mouse
- Western blot:Yes
- Gene ID:948
- Antigen synonyms:GP4|GP3B|FAT|GPIV|SCARB3|CHDS7|BDPLT10|PASIV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone: D-2712
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat