Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 755) [clone: 2G10]
Catalog # NOVUNB110-59722IR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:KIAA1524
- Clonality:Monoclonal
- Clone:2G10
- Conjugation:DyLight® 755
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Western blot:Yes
- Gene ID:57650
- Antigen synonyms:CIP2A|p90
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone: 2G10
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: