Anti-CDKN2A Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB110-59085B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2A (melanoma p16 inhibits CDK4)
- Antigen symbol:p14ARF
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:1029
- Antigen synonyms:ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human