Anti-TMED10 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB110-57586V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:transmembrane p24 trafficking protein 10
- Antigen symbol:TMED10
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Western blot:Yes
- Gene ID:10972
- Antigen synonyms:Tmp-21-I|S31III125|p23|TMP21|S31I125|P24(DELTA)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TMED10 (transmembrane emp24-like trafficking protein 10 (yeast))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: