Order Entry
France
ContactUsLinkComponent
Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 4H8H8E2]
Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 4H8H8E2]
Catalog # NOVUNB110-57307C
Supplier:  Novus Biologicals
Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 4H8H8E2]
Catalog # NOVUNB110-57307C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein A5
  • Antigen symbol:
    APOA5
  • Clonality:
    Monoclonal
  • Clone:
    4H8H8E2
  • Conjugation:
    DyLight® 650
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    116519
  • Antigen synonyms:
    MGC126836|FLJ97995|RAP3|APOAV|MGC126838
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: monoclonal
Clone: 4H8H8E2
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human