Anti-TARDBP Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNB110-55376C
- Antibody type:Primary
- Antigen name:TAR DNA binding protein
- Antigen symbol:TARDBP
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:23435
- Antigen synonyms:ALS10|TDP-43|RP4-635E18.2|TAR DNA-binding protein 43|TAR DNA-binding protein-43
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: