Anti-OPA1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB110-55290G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Optic Atrophy 1
- Antigen symbol:OPA1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4976
- Antigen synonyms:optic atrophy protein 1|mitochondrial dynamin-like 120 kDa protein|largeG|NTG|mitochondrial|MGM1|dynamin-like guanosine triphosphatase|dynamin-like 120 kDa protein|NPG|mitochondrial dynamin-like GTPase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: OPA1 (optic atrophy 1 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse, Rat