Order Entry
France
ContactUsLinkComponent
Anti-Smallpox A27L Rabbit Polyclonal Antibody
Anti-Smallpox A27L Rabbit Polyclonal Antibody
  NOVUNB100-56486
undefined
Anti-Smallpox A27L Rabbit Polyclonal Antibody
  NOVUNB100-56486

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Smallpox A27L
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: Smallpox A27L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Virus