Anti-RICTOR Rabbit Polyclonal Antibody
NOVUNB100-56427
- Antibody type:Primary
- Antigen name:RPTOR independent companion of MTOR, complex 2
- Antigen symbol:RICTOR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:253260
- Antigen synonyms:AVO3|PIA|hAVO3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: RICTOR (RPTOR independent companion of MTOR, complex 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human