Anti-SMAC/Diablo Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56165
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:SMAC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:56616
- Antigen synonyms:FLJ10537|SMAC|SMAC3|FLJ25049|DIABLO|DIABLO-S|0610041G12Rik
- Storage buffer:Whole antisera
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
The SMAC/Diablo Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMAC/Diablo. This antibody reacts with human, mouse, rat, gerbil. The SMAC/Diablo Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000
Type: Primary
Antigen: SMAC/Diablo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Gerbil