Anti-BFAR Rabbit Polyclonal Antibody
NOVUNB100-56093
- Antibody type:Primary
- Antigen name:bifunctional apoptosis regulator
- Antigen symbol:BFAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:51283
- Antigen synonyms:RNF47|BAR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Type: Primary
Antigen: BFAR (bifunctional apoptosis regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human