Order Entry
France
ContactUsLinkComponent
Anti-Separase Mouse Monoclonal Antibody [clone: XJ11-1B12]
Anti-Separase Mouse Monoclonal Antibody [clone: XJ11-1B12]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-Separase Mouse Monoclonal Antibody [clone: XJ11-1B12]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-439
  • NOVUNB100-439SS
  • Antigen symbol:
  • ESPL1
  • ESPL1
  • Antigen name:
  • Separase
  • Separase
  • Antigen synonyms:
  • SEPA|ESP1
  • SEPA|ESP1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • XJ11-1B12
  • XJ11-1B12
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 9700
  • 9700
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together