Order Entry
France
ContactUsLinkComponent
Anti-RPE65 Mouse Monoclonal Antibody [clone: 401.8B11.3D9]
Anti-RPE65 Mouse Monoclonal Antibody [clone: 401.8B11.3D9]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-RPE65 Mouse Monoclonal Antibody [clone: 401.8B11.3D9]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-355SS
  • NOVUNB100-355
  • Antigen symbol:
  • RPE65
  • RPE65
  • Antigen name:
  • Retinal Pigment Epithelium-specific 65 kDa protein
  • Retinal Pigment Epithelium-specific 65 kDa protein
  • Antigen synonyms:
  • sRPE65|all-trans-retinyl-palmitate hydrolase|retinol isomerase|retinoid isomerohydrolase|retinitis pigmentosa 20 (autosomal recessive)|retinal pigment epithelium-specific 65 kDa protein|RBP-binding membrane protein|p63|LCA2|RP20|rd12|mRPE65
  • sRPE65|all-trans-retinyl-palmitate hydrolase|retinol isomerase|retinoid isomerohydrolase|retinitis pigmentosa 20 (autosomal recessive)|retinal pigment epithelium-specific 65 kDa protein|RBP-binding membrane protein|p63|LCA2|RP20|rd12|mRPE65
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 401.8B11.3D9
  • 401.8B11.3D9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 6121
  • 6121
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together