Anti-SIRT6 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNB100-2524C
- Antibody type:Primary
- Antigen name:Sirtuin 6
- Antigen symbol:SIRT6
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:51548
- Antigen synonyms:SIR2-like protein 6|Mono-ADP-ribosyltransferase sirtuin-6|Silent information regulator 2-like protein 6.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human