Anti-CRYAB Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB100-2519G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Crystallin AB
- Antigen symbol:CRYAB
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:1410
- Antigen synonyms:HSPB5|CRYA2|CMD1II|CTPP2|HEL-S-101|CTRCT16|MFM2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse