Anti-PECAM1 Rat Monoclonal Antibody (DyLight® 755) [clone: MEC 7.46]
Catalog # NOVUNB100-1642IR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Platelet/endothelial cell adhesion molecule 1
- Antigen symbol:PECAM1
- Clonality:Monoclonal
- Clone:MEC 7.46
- Conjugation:DyLight® 755
- Flow cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Gene ID:5175
- Antigen synonyms:endoCAM|PECAM-1|GPIIA'|CD31|CD31/EndoCAM|PECA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEC 7.46
Conjugation: DyLight® 755
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse