Anti-CYP19A1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB100-1596V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:cytochrome P450, family 19, subfamily A, polypeptide 1
- Antigen symbol:CYP19A1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:1588
- Antigen synonyms:ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: