Anti-CYP19A1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB100-1596H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450 Family 19 Subfamily A Member 1
- Antigen symbol:CYP19A1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:1588
- Antigen synonyms:ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM, Aromatase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: