Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB100-134C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor)
- Antigen symbol:HIF1A
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Gel shift:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- ChIP:Yes
- Gene ID:3091
- Antigen synonyms:PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: