Order Entry
France
ContactUsLinkComponent
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-123SS
  • NOVUNB100-123
  • Antigen symbol:
  • HIF1A
  • HIF1A
  • Antigen name:
  • HIF1 alpha
  • HIF1 alpha
  • Antigen synonyms:
  • PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • H1alpha67
  • H1alpha67
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3091
  • 3091
  • Isotype:
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Gel shift:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together