Order Entry
France
ContactUsLinkComponent
 
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
 :  Biotium
 
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
 :  Biotium

 

  • Catalog No:
  • 20286-1
  • 20286.
  • Antigen symbol:
  • IgG
  • IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Reactivity:
  • Pig
  • Pig
  • Host:
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together