Order Entry
France
ContactUsLinkComponent
 
Anti-AGL/Alpha-glucosidase Rabbit Monoclonal Antibody [clone: EPR8880]
 :  Abcam
New Product
 
undefined
Anti-AGL/Alpha-glucosidase Rabbit Monoclonal Antibody [clone: EPR8880]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB133720-100
  • ABCAAB133720-1ML
  • ABCAAB133720-10
  • Antigen symbol:
  • AGL
  • Antigen name:
  • amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR8880
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

Frequently Bought Together