Order Entry
France
ContactUsLinkComponent
Anti-BACH1.3 Mouse Monoclonal Antibody [clone: OTI4E11]
Anti-BACH1.3 Mouse Monoclonal Antibody [clone: OTI4E11]
Catalog # ABCAAB128486-100
Supplier:  Abcam
undefined
Anti-BACH1.3 Mouse Monoclonal Antibody [clone: OTI4E11]
Catalog # ABCAAB128486-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transcription regulator protein BACH1
  • Antigen symbol:
    BACH1.3
  • Clonality:
    Monoclonal
  • Clone:
    4E11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    BACH1|BTB and CNC homolog 1|Transcription regulator protein BACH1|HA2303|BACH1.3
  • Storage buffer:
    Preservative: 0,02% Sodium azide
    Constituents: PBS, 50% Glycerol (glycerin, glycerine), 1% BSA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    WB
    Flow cytometry
    ImmunoChemistry
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BACH1.3
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human