Anti-Neurotensin Receptor 1 Mouse Monoclonal Antibody [clone: B-N6]
ABCAAB27380-1
: Abcam
- Antibody type:Primary
- Antigen name:Neurotensin Receptor 1
- Antigen symbol:NTSR1
- Clonality:Monoclonal
- Clone:B-N6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Neurotensin Receptor 1
Clonality: monoclonal
Clone: B-N6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human