Order Entry
France
ContactUsLinkComponent
Anti-Firefly Luciferase Mouse Monoclonal Antibody [clone: Luci17]
Anti-Firefly Luciferase Mouse Monoclonal Antibody [clone: Luci17]
Catalog # ABCAAB16466-100
Supplier:  Abcam
undefined
Anti-Firefly Luciferase Mouse Monoclonal Antibody [clone: Luci17]
Catalog # ABCAAB16466-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Firefly Luciferase
  • Clonality:
    Monoclonal
  • Clone:
    Luci17
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Firefly Luciferase
Clonality: monoclonal
Clone: Luci17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: