Anti-NLRP12 Mouse Monoclonal Antibody [clone: 4B9]
ABNOH00091662-M03
: Abnova
- Antibody type:Primary
- Antigen name:NACHT, LRR And PYD Domains-Containing protein 12
- Antigen symbol:NLRP12
- Clonality:Monoclonal
- Clone:4B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:91662
- Antigen synonyms:PAN6|RNO2|RNO|NALP12|PYPAF7|FCAS2|CLR19.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NLRP12
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human