Anti-REXO2 Mouse Monoclonal Antibody [clone: 2B5]
Catalog # ABNOH00025996-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RNA Exonuclease 2
- Antigen symbol:REXO2
- Clonality:Monoclonal
- Clone:2B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:25996
- Antigen synonyms:REX2|RFN|SFN|CGI-114
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: REXO2
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human