Order Entry
France
ContactUsLinkComponent
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 7B7]
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 7B7]
Catalog # ABNOH00023411-M01
Supplier:  Abnova
undefined
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 7B7]
Catalog # ABNOH00023411-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sirtuin 1
  • Antigen symbol:
    SIRT1
  • Clonality:
    Monoclonal
  • Clone:
    7B7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    23411
  • Antigen synonyms:
    NAD-dependent Deacetylase Sirtuin-1|SIR2L1|SIRT1|SIR2-like Protein 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SIRT1
Clonality: Monoclonal
Clone: 7B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human