Order Entry
France
ContactUsLinkComponent
Anti-LARS2 Mouse Monoclonal Antibody [clone: 3F12]
Anti-LARS2 Mouse Monoclonal Antibody [clone: 3F12]
Catalog # ABNOH00023395-M03
Supplier:  Abnova
Anti-LARS2 Mouse Monoclonal Antibody [clone: 3F12]
Catalog # ABNOH00023395-M03
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leucyl-tRNA Synthetase 2, mitochondrial
  • Antigen symbol:
    LARS2
  • Clonality:
    Monoclonal
  • Clone:
    3F12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    23395
  • Antigen synonyms:
    PRLTS4|LEURS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: LARS2
Clonality: Monoclonal
Clone: 3F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human