Anti-LILRA2 Mouse Monoclonal Antibody [clone: 3C7]
Catalog # ABNOH00011027-M17
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Leukocyte immunoglobulin-like Receptor subfamily A (with TM Domain) member 2
- Antigen symbol:LILRA2
- Clonality:Monoclonal
- Clone:3C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11027
- Antigen synonyms:LIR-7|CD85H|ILT1|LIR7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LILRA2
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human