Anti-IRX5 Mouse Monoclonal Antibody [clone: 6B5]
Catalog # ABNOH00010265-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Iroquois Homeobox Protein 5
- Antigen symbol:IRX5
- Clonality:Monoclonal
- Clone:6B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10265
- Antigen synonyms:HMMS|IRXB2|IRX-2a
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IRX5
Clonality: Monoclonal
Clone: 6B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human