Anti-ZBED1 Rabbit Polyclonal Antibody
Catalog # ABNOH00009189-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger, BED-type Containing 1
- Antigen symbol:ZBED1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9189
- Antigen synonyms:Zinc finger BED domain-containing protein 1|ALTE|Putative Ac-like transposable element|KIAA0785|DREF|hDREF|TRAMP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ZBED1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human