Anti-FADD Mouse Monoclonal Antibody [clone: 1D5]
Catalog # ABNOH00008772-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fas-associated protein with death domain
- Antigen symbol:FADD
- Clonality:Monoclonal
- Clone:1D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8772
- Antigen synonyms:FAS-associated death domain-containing protein|MORT1|MGC8528|GIG3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FADD
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human