Anti-WEE1 (WEE1 G2 checkpoint kinase) Mouse Polyclonal Antibody
Catalog # ABNOH00007465-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:WEE1 G2 checkpoint kinase
- Antigen symbol:WEE1 (WEE1 G2 checkpoint kinase)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7465
- Antigen synonyms:WEE1A|WEE1hu
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: WEE1 (WEE1 G2 checkpoint kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human